Pentafluorophenyl 2,4,6-trimethylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

043128-1G
101859-314EA 467.69 USD
101859-314
Pentafluorophenyl 2,4,6-trimethylbenzenesulfonate ≥95%
Pentafluorophenyl 2,4,6-trimethylbenzenesulphonate

Hazard: Irritant

Formula: C₁₅H₁₁F₅O₃S
MDL Number: MFCD05975138
CAS Number: 885950-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR