3-(2,6-Dimethylmorpholino)-1,1,1-trifluoro-2-propanol ≥95%
Supplier: Matrix Scientific
043050-500MG
043050-5G
101858-886EA
1109.23
USD
101858-886
101858-890
3-(2,6-Dimethylmorpholino)-1,1,1-trifluoro-2-propanol ≥95%
3-(2,6-Dimethylmorpholino)-1,1,1-trifluoro-2-propanol
Hazard: Irritant
Formula:
C₉H₁₆F₃NO₂ MW: 227.23 g/mol |
MDL Number:
MFCD03001226 CAS Number: 667938-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...