Ethyl-N-(2-cyano-3-ethoxyacryloyl)carbamate ≥95%

Supplier: Matrix Scientific

042918-1G
101858-218EA 698.46 USD
101858-218
Ethyl-N-(2-cyano-3-ethoxyacryloyl)carbamate ≥95%
Ethyl-N-(2-cyano-3-ethoxyacryloyl)carbamate

Hazard: Irritant

Formula: C₉H₁₂N₂O₄
MDL Number: MFCD00114882
CAS Number: 1187-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR