1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-2-ethylpiperidine ≥95%

Supplier: Matrix Scientific

042888-1G
101858-058EA 350.77 USD
101858-058
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-2-ethylpiperidine ≥95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-2-ethylpiperidine

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂S
MDL Number: MFCD08276942
CAS Number: 885950-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR