Pentafluorophenyl 3-(trifluoromethyl)benzenesulfonate ≥95%
Supplier: Matrix Scientific
042806-5G
042806-500MG
042806-1G
101857-636EA
1073.85
USD
101857-636
101857-634
101857-632
Pentafluorophenyl 3-(trifluoromethyl)benzenesulfonate ≥95%
Pentafluorophenyl 3-(trifluoromethyl)benzenesulphonate
Hazard: Irritant
Formula:
C₁₃H₄F₈O₃S |
MDL Number:
MFCD05975128 CAS Number: 885950-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...