2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)phenylglycine2-Amino-2-(3-(trifluoromethyl)phenyl)acetic acid

042734-1G
101857-250EA 420 USD
101857-250
2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid ≥95%
2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid

Hazard: Irritant

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD02662404
CAS Number: 242475-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR