3-Chloro-5-(trifluoromethyl)picolinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-5-(trifluoromethyl)pyridine-2-carbonitrile

042523-1G 042523-5G
101856-174EA 33.85 USD
101856-174 101856-176
3-Chloro-5-(trifluoromethyl)picolinonitrile ≥95%
3-Chloro-5-(trifluoromethyl)picolinonitrile

Hazard: Irritant

Formula: C₇H₂ClF₃N₂
MW: 206.5524 g/mol
MDL Number: MFCD00664066
CAS Number: 80194-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR