MATR042496

042496-1G
101856-036EA 143.69 USD
101856-036
MATR042496
Purity: 98%
CAS: 1189513-55-0
MDL Number: MFCD12827684
Molecular Formula: C6H3BrFNO2
Molecular Weight: 220
Hazard: Irritant-Harmful/Refrigerate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR