[(1-Methylpiperidin-4-yl)methyl]amine hydrochloride ≥97%

Supplier: Matrix Scientific

042491-1G 042491-5G
101856-010EA 127.69 USD
101856-010 101856-012
[(1-Methylpiperidin-4-yl)methyl]amine hydrochloride ≥97%
[(1-Methylpiperidin-4-yl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂
MDL Number: MFCD12827683
CAS Number: 1187582-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR