N-Methyl-L-proline ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Me-Pro-OH
, H-N-Me-Pro-OH
042486-5G
042486-1G
101855-992EA
47.69
USD
101855-992
101855-994
N-Methyl-L-proline ≥95%
N-Methyl-L-proline
Hazard: Irritant
Formula:
C₆H₁₁NO₂ MW: 129.159 g/mol |
MDL Number:
MFCD00011565 CAS Number: 475-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...