Boc-N-Me-4-methoxy-Phe-OH . DCHA ≥97%

Supplier: Matrix Scientific

Synonyms: Boc-Metyr(Me)-OH DCHA

042452-1G 042452-5G
101855-860EA 67.85 USD
101855-860 101855-862
Boc-N-Me-4-methoxy-Phe-OH . DCHA ≥97%
Boc-N-Me-4-methoxy-Phe-OH . DCHA

Hazard: Irritant

Formula: C₂₈H₄₆N₂O₅
MW: 490.69 g/mol
MDL Number: MFCD00153394
CAS Number: 135103-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR