Boc-N-Me-Val-OH
Supplier: Matrix Scientific
Synonyms:
N-(tert-Butoxycarbonyl)-N-methyl-L-valine
, Boc-N-Me-Val-OH
, N-Boc-N-methyl-L-valine
042444-1G
042444-5G
101855-830EA
304.62
USD
101855-830
101855-828
Boc-N-Me-Val-OH
Boc-N-Me-Val-OH
CAS: 45170-31-8
MDL Number: MFCD00038760
Molecular Formula: C11H21NO4
Molecular Weight: 231,29
Hazard: Irritant
Formula:
C₁₁H₂₁NO₄ MW: 231.292 g/mol |
MDL Number:
MFCD00038760 CAS Number: 45170-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...