Boc-Nα-methyl-L-tryptophan

Supplier: Matrix Scientific

042443-100MG 042443-1G
101855-824EA 230.77 USD
101855-824 101855-826
Boc-Nα-methyl-L-tryptophan
Boc-Nalpha-methyl-L-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MW: 318.38 g/mol
MDL Number: MFCD02682368
CAS Number: 141408-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR