Boc-N-methyl-D-leucine
Supplier: Matrix Scientific
Synonyms:
(R)-2-((tert-Butoxycarbonyl)(methyl)amino)-4-methylpentanoic acid
, Boc-N-Me-D-Leu-OH
042430-1G
042430-5G
101855-772EA
796.92
USD
101855-772
101855-770
Boc-N-methyl-D-leucine
Boc-N-methyl-D-leucine
CAS: 89536-84-5
MDL Number: MFCD00076953
Molecular Formula: C12H23NO4
Molecular Weight: 245,32
Hazard: Irritant
Formula:
C₁₂H₂₃NO₄ MW: 245.319 g/mol |
MDL Number:
MFCD00076953 CAS Number: 89536-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...