Fmoc-1-pyrrolidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

042410-1G 042410-250MG
101855-694EA 113.85 USD
101855-694 101855-696
Fmoc-1-pyrrolidine-3-carboxylic acid ≥95%
Fmoc-1-pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₉NO₄
MDL Number: MFCD02179123
CAS Number: 885951-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR