(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
1-(tert-Butoxycarbonyl)-L-pipecolic Acid
, Boc-HomoPro-OH
, Boc-Pip-OH
, 1-Boc-L-pipecolic Acid
, (S)-N-BOC-Piperidine-2-carboxylic acid
, (S)-1-Boc-2-piperidinecarboxylic Acid
, Boc-Homopro-OH
042389-5G
042389-25G
101855-614EA
52.31
USD
101855-614
101855-612
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol Melting Pt: 121…125 °C |
MDL Number:
MFCD00151904 CAS Number: 26250-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...