Fmoc-L-β-homoisoleucine

Supplier: Matrix Scientific

Synonyms: Fmoc-β-HomoIle-OH

042367-1G 042367-5G
101855-526EA 2401.54 USD
101855-526 101855-524
Fmoc-L-β-homoisoleucine
Fmoc-L-beta-homoisoleucine

Hazard: Irritant

Formula: C₂₂H₂₅NO₄
MW: 367.45 g/mol
MDL Number: MFCD01862850
CAS Number: 193954-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR