Fmoc-(S)-3-amino-4-(3-methylphenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(m-tolyl)butanoic acid
042350-250MG
042350-1G
101855-464EA
752.31
USD
101855-464
101855-462
Fmoc-(S)-3-amino-4-(3-methylphenyl)butyric acid
Fmoc-(S)-3-amino-4-(3-methylphenyl)butyric acid
Hazard: Irritant
Formula:
C₂₆H₂₅NO₄ MW: 415.49 g/mol |
MDL Number:
MFCD01861024 CAS Number: 270062-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...