Fmoc-(S)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Supplier: Matrix Scientific
042343-1G
042343-250MG
101855-434EA
412.31
USD
101855-434
101855-436
Fmoc-(S)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Fmoc-(S)-3-amino-4-(2-trifluoromethylphenyl)butyric acid
Hazard: Irritant
Formula:
C₂₆H₂₂F₃NO₄ |
MDL Number:
MFCD01861065 CAS Number: 270065-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...