Fmoc-(S)-3-amino-4-(2-chlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(2-chlorophenyl)butanoic acid

042336-250MG 042336-1G
101855-406EA 275.38 USD
101855-406 101855-408
Fmoc-(S)-3-amino-4-(2-chlorophenyl)butyric acid
Fmoc-(S)-3-amino-4-(2-chlorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂ClNO₄
MW: 435.91 g/mol
MDL Number: MFCD01860997
CAS Number: 270596-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR