Fmoc-(R)-3-amino-4-pentafluorophenylbutyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(perfluorophenyl)butanoic acid
042327-250MG
042327-1G
101855-372EA
841.8
USD
101855-372
101855-370
Fmoc-(R)-3-amino-4-pentafluorophenylbutyric acid
Fmoc-(R)-3-amino-4-pentafluorophenylbutyric acid
Hazard: Irritant
Formula:
C₂₅H₁₈F₅NO₄ MW: 491.42 g/mol |
MDL Number:
MFCD01860941 CAS Number: 269398-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...