Fmoc-(R)-3-amino-4-(4-iodophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-iodophenyl)butanoic acid

042321-250MG 042321-1G
101855-346EA 275.38 USD
101855-346 101855-348
Fmoc-(R)-3-amino-4-(4-iodophenyl)butyric acid
Fmoc-(R)-3-amino-4-(4-iodophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂INO₄
MW: 527.36 g/mol
MDL Number: MFCD01860964
CAS Number: 269396-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR