Fmoc-(R)-3-amino-4-(4-fluorophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-fluorophenyl)butanoic acid
042320-1G
042320-250MG
101855-342EA
275.38
USD
101855-342
101855-344
Fmoc-(R)-3-amino-4-(4-fluorophenyl)butyric acid
Fmoc-(R)-3-amino-4-(4-fluorophenyl)butyric acid
Hazard: Irritant
Formula:
C₂₅H₂₂FNO₄ MW: 419.46 g/mol |
MDL Number:
MFCD01860915 CAS Number: 331763-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...