Fmoc-(R)-3-amino-4-(2-naphthyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(naphthalen-2-yl)butanoic acid
042304-1G
042304-250MG
101855-278EA
275.38
USD
101855-278
101855-280
Fmoc-(R)-3-amino-4-(2-naphthyl)butyric acid
Fmoc-(R)-3-amino-4-(2-naphthyl)butyric acid
Hazard: Irritant
Formula:
C₂₉H₂₅NO₄ MW: 451.53 g/mol |
MDL Number:
MFCD01860938 CAS Number: 269398-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...