Fmoc-(R)-3-amino-4-(2-furyl)butyric acid
Supplier: Matrix Scientific
042302-1G
042302-250MG
101855-272EA
1353.85
USD
101855-272
101855-270
Fmoc-(R)-3-amino-4-(2-furyl)butyric acid
Fmoc-(R)-3-amino-4-(2-furyl)butyric acid
Hazard: Irritant
Formula:
C₂₃H₂₁NO₅ |
MDL Number:
MFCD01860994 CAS Number: 270596-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...