Boc-L-β-homotryptophan
Supplier: Matrix Scientific
Synonyms:
(S)-3-((tert-Butoxycarbonyl)amino)-4-(1H-indol-3-yl)butanoic acid
042288-1G
042288-250MG
101855-214EA
238.46
USD
101855-214
101855-216
Boc-L-β-homotryptophan
Boc-L-beta-homotryptophan
Hazard: Irritant
Formula:
C₁₇H₂₂N₂O₄ MW: 318.38 g/mol |
MDL Number:
MFCD01862938 CAS Number: 229639-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...