Boc-L-β-homoalanine

Supplier: Matrix Scientific

Synonyms: Boc-β-HoAla-OHBoc-β-HomoAla-OH

042280-5G 042280-1G
101855-184EA 206.15 USD
101855-184 101855-182
Boc-L-β-homoalanine
Boc-L-beta-homoalanine

Hazard: Irritant

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
MDL Number: MFCD00270345
CAS Number: 158851-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR