Boc-DL-β-leucine

Supplier: Matrix Scientific

Synonyms: 3-(Boc-amino)-4-methylpentanoic acid

042279-1G
101855-178EA 1976.92 USD
101855-178
Boc-DL-β-leucine
Boc-DL-beta-leucine

Hazard: Irritant

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
MDL Number: MFCD02258127
CAS Number: 248924-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR