Boc-3-aminoadipic acid

Supplier: Matrix Scientific

042276-250MG 042276-1G
101855-166EA 400 USD
101855-166 101855-168
Boc-3-aminoadipic acid
Boc-3-aminoadipic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₆
MDL Number: MFCD02682491
CAS Number: 1185301-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR