3-(Boc-amino)-3-(4-chlorophenyl)propionic acid

Supplier: Matrix Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid

042274-100MG 042274-500MG
101855-158EA 300 USD
101855-158 101855-160
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
MDL Number: MFCD02090708
CAS Number: 284493-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR