Boc-(S)-3-amino-6-phenyl-5-hexenoic acid
Supplier: Matrix Scientific
Synonyms:
(S)-3-((tert-Butoxycarbonyl)amino)-6-phenylhex-5-enoic acid
042268-1G
042268-250MG
101855-138EA
412.31
USD
101855-138
101855-140
Boc-(S)-3-amino-6-phenyl-5-hexenoic acid
Boc-(S)-3-amino-6-phenyl-5-hexenoic acid
Hazard: Irritant
Formula:
C₁₇H₂₃NO₄ MW: 305.38 g/mol |
MDL Number:
MFCD01861004 CAS Number: 270596-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...