Boc-(S)-3-amino-5-hexynoic acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)hex-5-ynoic acid

042266-250MG 042266-1G
101855-132EA 575 USD
101855-132 101855-130
Boc-(S)-3-amino-5-hexynoic acid
Boc-(S)-3-amino-5-hexynoic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD01861007
CAS Number: 270596-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR