Boc-(S)-3-amino-4-(4-pyridyl)butyric acid

Supplier: Matrix Scientific

042262-250MG 042262-1G
101855-116EA 1376.92 USD
101855-116 101855-114
Boc-(S)-3-amino-4-(4-pyridyl)butyric acid
Boc-(S)-3-amino-4-(4-pyridyl)butyric acid

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₄
MW: 280.33 g/mol
MDL Number: MFCD01076288
CAS Number: 219297-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR