Boc-(S)-3-amino-4-(4-iodophenyl)butyric acid
Supplier: Matrix Scientific
Synonyms:
(S)-3-((tert-Butoxycarbonyl)amino)-4-(4-iodophenyl)butanoic acid
042259-1G
042259-250MG
101855-104EA
752.31
USD
101855-104
101855-102
Boc-(S)-3-amino-4-(4-iodophenyl)butyric acid
Boc-(S)-3-amino-4-(4-iodophenyl)butyric acid
Hazard: Irritant
Formula:
C₁₅H₂₀INO₄ MW: 405.24 g/mol |
MDL Number:
MFCD01861061 CAS Number: 270065-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...