Boc-(S)-3-amino-4-(3-thienyl)butyric acid

Supplier: Matrix Scientific

042253-250MG 042253-1G
101855-078EA 412.31 USD
101855-078 101855-080
Boc-(S)-3-amino-4-(3-thienyl)butyric acid
Boc-(S)-3-amino-4-(3-thienyl)butyric acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₄S
MW: 285.36 g/mol
MDL Number: MFCD01861084
CAS Number: 270263-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR