Boc-(R)-3-amino-4-pentafluorophenylbutyric acid
Supplier: Matrix Scientific
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-4-(perfluorophenyl)butanoic acid
042228-1G
042228-250MG
101854-980EA
1126.15
USD
101854-980
101854-978
Boc-(R)-3-amino-4-pentafluorophenylbutyric acid
Boc-(R)-3-amino-4-pentafluorophenylbutyric acid
Hazard: Irritant
Formula:
C₁₅H₁₆F₅NO₄ MW: 369.29 g/mol |
MDL Number:
MFCD01860940 CAS Number: 269398-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...