Boc-(R)-3-amino-4-(4-cyanophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(4-cyanophenyl)butanoic acid

042220-250MG
101854-946EA 279.45 USD
101854-946
Boc-(R)-3-amino-4-(4-cyanophenyl)butyric acid
Boc-(R)-3-amino-4-(4-cyanophenyl)butyric acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₄
MW: 304.35 g/mol
MDL Number: MFCD01860981
CAS Number: 269726-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR