Boc-(R)-3-amino-4-(3-thienyl)butyric acid

Supplier: Matrix Scientific

042216-250MG 042216-1G
101854-930EA 412.31 USD
101854-930 101854-932
Boc-(R)-3-amino-4-(3-thienyl)butyric acid
Boc-(R)-3-amino-4-(3-thienyl)butyric acid

Hazard: Irritant

Formula: C₁₃H₁₉NO₄S
MW: 285.36 g/mol
MDL Number: MFCD01860987
CAS Number: 269726-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR