Boc-(R)-3-amino-4-(3-chlorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(3-chlorophenyl)butanoic acid

042211-1G 042211-250MG
101854-910EA 275.38 USD
101854-910 101854-912
Boc-(R)-3-amino-4-(3-chlorophenyl)butyric acid
Boc-(R)-3-amino-4-(3-chlorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₅H₂₀ClNO₄
MW: 313.78 g/mol
MDL Number: MFCD01860901
CAS Number: 331763-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR