4-(Benzyloxy)-3-ethoxybenzoic acid

Supplier: Matrix Scientific

040097-500MG
101848-832EA 144.9 USD
101848-832
4-(Benzyloxy)-3-ethoxybenzoic acid
4-(Benzyloxy)-3-ethoxybenzoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₄
MDL Number: MFCD05039683
CAS Number: 52009-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR