3-[(2-Methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040095-500MG
101848-828EA 193.85 USD
101848-828
3-[(2-Methylbenzyl)oxy]benzoic acid
3-[(2-Methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD08443025
CAS Number: 926228-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR