3-[(2-Fluorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040092-500MG
101848-822EA 144.9 USD
101848-822
3-[(2-Fluorobenzyl)oxy]benzoic acid
3-[(2-Fluorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁FO₃
MDL Number: MFCD09705826
CAS Number: 938138-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR