3-[(4-Bromobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040091-500MG
101848-820EA 193.85 USD
101848-820
3-[(4-Bromobenzyl)oxy]benzoic acid
3-[(4-Bromobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁BrO₃
MDL Number: MFCD09705998
CAS Number: 855749-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR