5-Bromo-2-[(2-chloro-6-fluorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040089-500MG
101848-816EA 144.9 USD
101848-816
5-Bromo-2-[(2-chloro-6-fluorobenzyl)oxy]benzoic acid
5-Bromo-2-[(2-chloro-6-fluorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₉BrClFO₃
MDL Number: MFCD09708239
CAS Number: 938257-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR