2-[(3,4-Dichlorobenzyl)oxy]-3-methoxybenzoic acid

Supplier: Matrix Scientific

040087-500MG
101848-812EA 144.9 USD
101848-812
2-[(3,4-Dichlorobenzyl)oxy]-3-methoxybenzoic acid
2-[(3,4-Dichlorobenzyl)oxy]-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₄
MDL Number: MFCD09721279
CAS Number: 938274-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR