4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid

Supplier: Matrix Scientific

040085-500MG
101848-808EA 123.05 USD
101848-808
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid
4-[(3,4-Dichlorobenzyl)oxy]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂O₃
MW: 297.14 g/mol
MDL Number: MFCD06200988
CAS Number: 56442-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR