4-[(3-Chlorobenzyl)oxy]-3-ethoxybenzoic acid

Supplier: Matrix Scientific

040082-500MG
101848-802EA 193.85 USD
101848-802
4-[(3-Chlorobenzyl)oxy]-3-ethoxybenzoic acid
4-[(3-Chlorobenzyl)oxy]-3-ethoxybenzoic acid

Hazard: Irritant

Formula: C₁₆H₁₅ClO₄
MDL Number: MFCD09718403
CAS Number: 938375-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR