3-Ethoxy-4-[(3-methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040081-500MG
101848-800EA 193.85 USD
101848-800
3-Ethoxy-4-[(3-methylbenzyl)oxy]benzoic acid
3-Ethoxy-4-[(3-methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₇H₁₈O₄
MDL Number: MFCD09709235
CAS Number: 938271-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR