2-[(2,6-Dichlorobenzyl)oxy]-3-methoxybenzoic acid

Supplier: Matrix Scientific

040072-500MG
101848-782EA 144.9 USD
101848-782
2-[(2,6-Dichlorobenzyl)oxy]-3-methoxybenzoic acid
2-[(2,6-Dichlorobenzyl)oxy]-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₄
MDL Number: MFCD09721384
CAS Number: 938250-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR