5-Bromo-2-[(3-methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040071-500MG
101848-780EA 144.9 USD
101848-780
5-Bromo-2-[(3-methylbenzyl)oxy]benzoic acid
5-Bromo-2-[(3-methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃BrO₃
MDL Number: MFCD09720743
CAS Number: 938241-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR